Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025349.2 | NP_079625.1 | 2 | 96 | ||
GenBank mRNA | AK003062.1 | - | 2 | 84 | ||
AK007462.1 | - | 2 | 93 | |||
AK009317.1 | - | 2 | 83 | |||
AK012446.1 | - | 2 | 93 | |||
AK013355.1 | - | 2 | 97 | |||
BC027511.1 | - | 2 | 86 | 1382565 | ||
BC081444.1 | - | 1 | 79 | 5711629 |