Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025356.4 | NP_079632.1 | 4 | 348 | ||
GenBank mRNA | AK003163.1 | - | 4 | 299 | ||
AK004001.1 | - | 4 | 358 | |||
AK009276.1 | - | 4 | 290 | |||
AK009936.1 | - | 4 | 287 | |||
AK032885.1 | - | 4 | 358 | |||
AK049906.1 | - | 4 | 349 | |||
AK077666.1 | - | 4 | 408 | |||
AK088448.1 | - | 4 | 318 | |||
AK151295.1 | - | 4 | 462 | |||
AK151594.1 | - | 4 | 369 | |||
AK152231.1 | - | 4 | 369 | |||
AK152541.1 | - | 4 | 342 | |||
AK153100.1 | - | 4 | 462 | |||
AK153111.1 | - | 4 | 370 | |||
AK153410.1 | - | 4 | 369 | |||
AK153423.1 | - | 4 | 408 | |||
AK159125.1 | - | 4 | 408 | |||
AK159546.1 | - | 4 | 344 | |||
AK159963.1 | - | 4 | 342 | |||
AK165235.1 | - | 3 | 589 | |||
AK167077.1 | - | 4 | 358 | |||
AK167138.1 | - | 4 | 311 | |||
AK167650.1 | - | 4 | 346 | |||
AK167801.1 | - | 4 | 370 | |||
AK169177.1 | - | 3 | 720 | |||
AK208479.1 | - | 3 | 129 | |||
BC023266.1 | - | 3 | 113 | |||
BC057941.1 | - | 4 | 329 | 5323636 | ||
BC098220.1 | - | 4 | 363 | 963757 |