Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025356.4 | NP_079632.1 | 8 | 1106 | ||
GenBank mRNA | AK003163.1 | - | 8 | 1057 | ||
AK004001.1 | - | 8 | 1116 | |||
AK009276.1 | - | 8 | 1048 | |||
AK014344.1 | - | 2 | 587 | |||
AK032885.1 | - | 8 | 1116 | |||
AK049906.1 | - | 8 | 1107 | |||
AK077666.1 | - | 8 | 1166 | |||
AK088448.1 | - | 8 | 1076 | |||
AK151295.1 | - | 8 | 1220 | |||
AK151594.1 | - | 8 | 1127 | |||
AK152231.1 | - | 8 | 1127 | |||
AK152541.1 | - | 8 | 1100 | |||
AK153100.1 | - | 8 | 1220 | |||
AK153111.1 | - | 8 | 1128 | |||
AK153410.1 | - | 8 | 1127 | |||
AK153423.1 | - | 8 | 1166 | |||
AK159125.1 | - | 8 | 1166 | |||
AK159546.1 | - | 8 | 1102 | |||
AK159963.1 | - | 8 | 1100 | |||
AK165235.1 | - | 7 | 1347 | |||
AK167077.1 | - | 8 | 1116 | |||
AK167138.1 | - | 8 | 1069 | |||
AK167650.1 | - | 8 | 1104 | |||
AK167801.1 | - | 8 | 1128 | |||
AK169177.1 | - | 7 | 1479 | |||
BC023266.1 | - | 7 | 871 | |||
BC057941.1 | - | 8 | 1087 | 5323636 | ||
BC098220.1 | - | 8 | 1121 | 963757 |