Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252591.2 | NP_001239520.1 | 5 | 850 | ||
GenBank mRNA | AJ245772.1 | - | 6 | 740 | ||
AK010172.1 | - | 6 | 732 | |||
AY026524.1 | - | 6 | 731 | |||
BC061051.1 | - | 6 | 646 | 30300606 | ||
BC132358.1 | - | 6 | 560 | 40130635 | ||
BC138801.1 | - | 6 | 560 | 8861823 |