Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252591.2 | NP_001239520.1 | 5 | 922 | ||
GenBank mRNA | AF364070.1 | - | 6 | 796 | ||
AJ245772.1 | - | 6 | 813 | |||
AK003105.1 | - | 6 | 805 | |||
AK010172.1 | - | 6 | 804 | |||
BC061051.1 | - | 6 | 718 | 30300606 | ||
BC132358.1 | - | 6 | 632 | 40130635 | ||
BC138801.1 | - | 6 | 632 | 8861823 |