Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025364.2 | NP_079640.1 | 6 | 498 | ||
GenBank mRNA | AK003453.1 | - | 6 | 350 | ||
AK018773.1 | - | 6 | 361 | |||
AK088266.1 | - | 6 | 347 | |||
AK146248.1 | - | 6 | 376 | |||
BC027510.1 | - | 6 | 337 | 1382354 | ||
BC094393.1 | - | 6 | 334 | |||
BC157909.1 | - | 6 | 362 | 9088111 | ||
BC158023.1 | - | 6 | 362 | 9007329 | ||
BC172063.1 | - | 6 | 359 | 9054349 |