Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025383.3 | NP_079659.1 | 8 | 1124 | ||
GenBank mRNA | AK003463.1 | - | 8 | 1090 | ||
AK009714.1 | - | 4 | 669 | |||
AK145874.1 | - | 8 | 1113 | |||
AK155871.1 | - | 8 | 1112 | |||
AK156566.1 | - | 8 | 1099 | |||
AK170987.1 | - | 8 | 1107 | |||
AK172538.1 | - | 8 | 1093 | |||
AK182189.1 | - | 1 | 71 | |||
AK183675.1 | - | 1 | 301 | |||
AK185901.1 | - | 1 | 47 | |||
AK186809.1 | - | 1 | 74 | |||
AK193216.1 | - | 1 | 299 | |||
AK207134.1 | - | 1 | 46 | |||
AK213883.1 | - | 1 | 37 | |||
AK218072.1 | - | 1 | 57 | |||
AK218317.1 | - | 1 | 63 | |||
AK219572.1 | - | 1 | 303 | |||
BC037069.1 | - | 8 | 1121 | 5002765 |