Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025415.3 | NP_079691.1 | 3 | 566 | ||
GenBank mRNA | AK002451.1 | - | 3 | 474 | ||
AK004163.1 | - | 3 | 474 | |||
AK077690.1 | - | 3 | 376 | |||
AK166688.1 | - | 3 | 483 | |||
BC022647.1 | - | 3 | 473 | 4216297 | ||
BC094919.1 | - | 3 | 442 | 6431137 |