Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025416.3 | NP_079692.1 | 6 | 1025 | ||
GenBank mRNA | AK003517.1 | - | 4 | 881 | ||
AK004155.1 | - | 6 | 1026 | |||
AK009228.1 | - | 6 | 1024 | |||
BC046437.1 | - | 6 | 1007 | 3969280 |