Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025424.2 | NP_079700.1 | 4 | 468 | ||
GenBank mRNA | AB126218.1 | - | 4 | 442 | ||
AK002413.1 | - | 4 | 469 | |||
AK004329.1 | - | 4 | 457 | |||
AK009304.1 | - | 4 | 459 | |||
AK020403.1 | - | 4 | 450 | |||
AK155319.1 | - | 4 | 467 | |||
BC048464.1 | - | 4 | 456 | 6596336 |