Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025429.2 | NP_079705.2 | 7 | 1301 | ||
GenBank mRNA | AK004500.1 | - | 7 | 1303 | ||
AK160167.1 | - | 7 | 1297 | |||
AK160456.1 | - | 1 | 269 | |||
BC104333.2 | - | 6 | 1268 | 40045808 |