Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025429.2 | NP_079705.2 | 7 | 1704 | ||
GenBank mRNA | AK004500.1 | - | 7 | 1706 | ||
AK160167.1 | - | 7 | 1700 | |||
AK160456.1 | - | 1 | 672 |