Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025430.3 | NP_079706.1 | 4 | 577 | ||
GenBank mRNA | AK004385.1 | - | 4 | 570 | ||
AK010223.1 | - | 4 | 566 | |||
AK010369.1 | - | 4 | 563 | |||
AK012807.1 | - | 4 | 564 | |||
AK039187.1 | - | 4 | 557 | |||
BB615775.1 | - | 4 | 577 | |||
BC028750.1 | - | 4 | 569 | 1245736 |