Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025432.3 | NP_079708.2 | 4 | 461 | ||
GenBank mRNA | AK002658.1 | - | 5 | 532 | ||
AK004389.1 | - | 486 | ||||
AK007889.1 | - | 4 | 462 | |||
AK007951.1 | - | 396 | ||||
AK008564.1 | - | 5 | 511 | |||
AK077643.1 | - | 487 | ||||
AY318751.1 | - | 519 | ||||
BC034845.1 | - | 496 | 1528500 | |||
BC061087.1 | - | 531 | 6589013 | |||
BC099551.1 | - | 503 | 30252145 |