Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002658.1 | - | 5 | 1070 | ||
AK004389.1 | - | 1024 | ||||
AK007951.1 | - | 934 | ||||
AK077643.1 | - | 1024 | ||||
AY318751.1 | - | 1057 | ||||
BC034845.1 | - | 1034 | 1528500 | |||
BC061087.1 | - | 1069 | 6589013 | |||
BC099551.1 | - | 1041 | 30252145 |