Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145645.2 | NP_001139117.1 | 2 | 642 | ||
NM_006573.4 | NP_006564.1 | 2 | 642 | |||
GenBank mRNA | AF116456.1 | - | 2 | 493 | ||
AF132600.1 | - | 2 | 511 | |||
AF134715.1 | - | 2 | 519 | |||
AF136293.1 | - | 2 | 375 | |||
AF186114.1 | - | 2 | 532 | |||
AK309629.1 | - | 3 | 412 | |||
AY129225.1 | - | 2 | 375 | |||
AY129226.1 | - | 2 | 144 | |||
AY129227.1 | - | 1 | 42 | |||
AY302751.1 | - | 2 | 375 | |||
AY358881.1 | - | 2 | 544 | |||
BC020674.1 | - | 2 | 554 | 4719315 | ||
BI818459.1 | - | 2 | 642 | |||
CR541818.1 | - | 2 | 375 | |||
HM636064.1 | - | 2 | 375 |