Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160145.1 | NP_001153617.1 | 6 | 1085 | ||
NM_001160146.1 | NP_001153618.1 | 6 | 981 | |||
NM_025439.3 | NP_079715.1 | 6 | 950 | |||
GenBank mRNA | AK134909.1 | - | 6 | 915 | ||
AK154423.1 | - | 6 | 937 | |||
AK186635.1 | - | 1 | 339 | |||
BC016524.1 | - | 6 | 924 | 3594475 | ||
BC020291.1 | - | 5 | 902 | |||
BC048017.1 | - | 6 | 904 |