Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025442.3 | NP_079718.1 | 2 | 251 | ||
GenBank mRNA | AK003818.1 | - | 2 | 232 | ||
AK005296.1 | - | 2 | 252 | |||
AK011159.1 | - | 2 | 220 | |||
AK030567.1 | - | 2 | 224 | |||
AK161187.1 | - | 232 | ||||
BC027160.1 | - | 2 | 211 | 4946430 |