Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025445.4 | NP_079721.2 | 16 | 2491 | ||
GenBank mRNA | AK002400.1 | - | 3 | 1060 | ||
AK007732.1 | - | 16 | 2411 | |||
AK028990.1 | - | 16 | 2492 | |||
AK075788.1 | - | 16 | 2477 | |||
AK169107.1 | - | 16 | 2398 | |||
BC004081.1 | - | 16 | 2427 | 3592719 | ||
BC060369.1 | - | 16 | 2415 | 6310619 |