Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 1, 2 | 157 | ||
GenBank mRNA | AK007759.1 | - | 1, 2 | 108 | ||
AK011544.1 | - | 1, 2 | 114 | |||
AK015145.1 | - | 1, 2 | 107 | |||
AK078186.1 | - | 1, 2 | 109 | |||
AK078401.1 | - | 1, 2 | 131 | |||
AK146188.1 | - | 1, 2 | 128 | |||
AK161333.1 | - | 1, 2 | 109 | |||
BC019799.1 | - | 1, 2 | 130 | 5053500 | ||
BF608861.1 | - | 1, 2 | 168 |