Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 12 | 1662 | ||
GenBank mRNA | AK011544.1 | - | 12 | 1619 | ||
AK015145.1 | - | 12 | 1611 | |||
AK078186.1 | - | 10 | 2456 | |||
AK146188.1 | - | 12 | 1633 | |||
AK161333.1 | - | 12 | 1614 |