Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 4 | 1726 | ||
GenBank mRNA | AK082374.1 | - | 4 | 1728 | ||
AK082512.1 | - | 168 |