Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025462.2 | NP_079738.2 | 2 | 452 | ||
NM_177940.1 | NP_808809.1 | 2 | 452 | |||
NM_177941.1 | NP_808810.1 | 2 | 452 | |||
GenBank mRNA | AF489569.1 | - | 2 | 271 | ||
AF489570.1 | - | 2 | 271 | |||
AK007407.1 | - | 2 | 461 | |||
AK007791.1 | - | 2 | 454 | |||
BC115541.2 | - | 2 | 454 | |||
BC115542.1 | - | 2 | 454 |