Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025462.2 | NP_079738.2 | 3 | 628 | ||
GenBank mRNA | AK007407.1 | - | 3 | 637 | ||
AK007791.1 | - | 3 | 630 | |||
BC115541.2 | - | 3 | 630 | |||
BC115542.1 | - | 2 | 1110 |