Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025473.3 | NP_079749.1 | 9 | 1705 | ||
GenBank mRNA | AK003680.1 | - | 1 | 651 | ||
AK007715.1 | - | 9 | 1706 | |||
BC011237.1 | - | 9 | 1703 | |||
BC054411.1 | - | 9 | 1379 | |||
BC069871.1 | - | 9 | 1841 | 6467541 |