Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdrt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025496.1 | NP_079772.1 | 2 | 260 | ||
GenBank mRNA | AK006125.1 | - | 2 | 260 | ||
AK006342.1 | - | 2 | 240 | |||
BC048461.1 | - | 2 | 227 | 6771281 |