Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256725.1 | NP_001243654.1 | 5 | 805 | ||
NM_001256726.1 | NP_001243655.1 | 5 | 805 | |||
NM_001256727.1 | NP_001243656.1 | 5 | 690 | |||
NM_001256728.1 | NP_001243657.1 | 4 | 615 | |||
NM_006590.3 | NP_006581.2 | 5 | 805 | |||
NR_046347.1 | 5 | 693 | ||||
XM_006711922.1 | XP_006711985.1 | 5 | 673 | |||
XM_006711923.2 | XP_006711986.1 | 5 | 752 | |||
XM_011532487.1 | XP_011530789.1 | 5 | 758 | |||
XM_011532488.1 | XP_011530790.1 | 5 | 691 | |||
XM_017003182.1 | XP_016858671.1 | 5 | 762 | |||
GenBank mRNA | AF130096.1 | - | 4 | 394 | ||
AF132955.1 | - | 5 | 710 | |||
AF353989.1 | - | 5 | 712 | |||
AK001047.1 | - | 5 | 703 | |||
AK001772.1 | - | 1 | 98 | |||
AK056334.1 | - | 5 | 674 | |||
AK129872.1 | - | 6 | 686 | |||
AK222852.1 | - | 5 | 708 | |||
AK289451.1 | - | 5 | 759 | |||
AK295257.1 | - | 4 | 613 | |||
AK302236.1 | - | 5 | 667 | |||
AK309485.1 | - | 6 | 608 | |||
AL117416.1 | - | 3 | 372 | |||
AL519827.3 | - | 5 | 755 | |||
BC001384.2 | - | 5 | 694 | |||
BC067273.1 | - | 5 | 733 | 6503509 | ||
BX462623.2 | - | 5 | 738 |