Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025519.2 | NP_079795.1 | 5 | 938 | ||
GenBank mRNA | AK009273.1 | - | 5 | 939 | ||
AK160189.1 | - | 5 | 932 | |||
BC115983.1 | - | 5 | 928 | 40105850 | ||
BC117538.1 | - | 5 | 928 | 40105849 |