Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006597.5 | NP_006588.1 | 3 | 636 | ||
NM_153201.3 | NP_694881.1 | 3 | 636 | |||
XM_011542798.1 | XP_011541100.1 | 3 | 384 | |||
GenBank mRNA | AB034951.1 | - | 3 | 400 | ||
AB073886.1 | - | 2 | 314 | |||
AF352832.1 | - | 3 | 423 | |||
AK096100.1 | - | 3 | 362 | |||
AK129885.1 | - | 3 | 436 | |||
AK222628.1 | - | 3 | 434 | |||
AK222785.1 | - | 3 | 434 | |||
AK300404.1 | - | 3 | 510 | |||
AK310467.1 | - | 3 | 508 | |||
BC007276.1 | - | 2 | 184 | |||
BC016179.1 | - | 3 | 417 | 3920744 | ||
BC016660.1 | - | 3 | 417 | 3923981 | ||
BC019816.2 | - | 3 | 417 | 2899894 | ||
EU668349.1 | - | 3 | 417 | |||
FJ224294.1 | - | 3 | 437 |