Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025523.1 | NP_079799.1 | 4 | 333 | ||
GenBank mRNA | AK003365.1 | - | 4 | 319 | ||
AK009393.1 | - | 4 | 333 | |||
AK018705.1 | - | 4 | 317 | |||
AK028011.1 | - | 317 | ||||
BC051933.1 | - | 4 | 314 | |||
BC092141.1 | - | 4 | 325 | 6814605 |