Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025534.2 | NP_079810.2 | 3 | 457 | ||
GenBank mRNA | AK009786.1 | - | 2 | 357 | ||
AK009932.1 | - | 2 | 371 | |||
AK141751.1 | - | 3 | 455 | |||
AK166406.1 | - | 2 | 397 | |||
BC057326.1 | - | 3 | 457 | 6826080 | ||
BC098496.1 | - | 2 | 384 | 30089818 |