Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025550.2 | NP_079826.2 | 6, 7 | 1025 | ||
GenBank mRNA | AJ252147.1 | - | 6, 7 | 1061 | ||
AK002343.1 | - | 6, 7 | 1061 | |||
AK010287.1 | - | 6, 7 | 1058 | |||
AK178742.1 | - | 3, 4 | 304 | |||
AK206699.1 | - | 1, 2 | 145 | |||
BC006869.1 | - | 6, 7 | 1025 | 2646325 |