Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025551.3 | NP_079827.2 | 4 | 351 | ||
GenBank mRNA | AK003131.1 | - | 4 | 324 | ||
AK003322.1 | - | 4 | 324 | |||
AK008876.1 | - | 4 | 324 | |||
AK010460.1 | - | 4 | 352 | |||
AK032774.1 | - | 4 | 325 | |||
AK134765.1 | - | 4 | 324 | |||
AK153240.1 | - | 4 | 324 | |||
AK167943.1 | - | 4 | 317 | |||
BC004633.1 | - | 1781 | ||||
BC086922.1 | - | 4 | 356 | 6751147 |