Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025558.5 | NP_079834.2 | 5 | 878 | ||
GenBank mRNA | AK003333.2 | - | 5 | 879 | ||
AK003815.1 | - | 5 | 868 | |||
AK007780.1 | - | 898 | ||||
AK052810.1 | - | 5 | 879 | |||
AK083359.1 | - | 5 | 724 | |||
AK088970.1 | - | 5 | 880 | |||
AK165543.1 | - | 5 | 882 | |||
AK166544.1 | - | 5 | 856 | |||
AK168422.1 | - | 5 | 868 | |||
BC046479.1 | - | 5 | 841 | |||
BC046810.1 | - | 5 | 844 | |||
BC048147.1 | - | 5 | 868 | |||
BC054749.1 | - | 5 | 899 | 6410090 | ||
BC058812.1 | - | 5 | 841 | 6530835 | ||
BC062980.1 | - | 5 | 890 | 6842939 |