Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006612.5 | NP_006603.2 | 6, 7 | 794 | ||
XM_005256424.2 | XP_005256481.1 | 7, 8 | 785 | |||
GenBank mRNA | AB014606.2 | - | 6, 7 | 790 | ||
AK307401.1 | - | 6, 7 | 637 | |||
BC034993.1 | - | 6, 7 | 768 | 5266798 | ||
BM458656.1 | - | 6, 7 | 795 | |||
U91329.1 | - | 5, 6 | 550 |