Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025566.3 | NP_079842.2 | 2 | 939 | ||
GenBank mRNA | AK004949.1 | - | 2 | 870 | ||
AK011245.1 | - | 2 | 901 | |||
AK090348.1 | - | 2 | 941 | |||
AK136962.1 | - | 2 | 901 | |||
AK150678.1 | - | 2 | 930 | |||
AK153177.1 | - | 2 | 925 | |||
BC032199.1 | - | 2 | 868 | 5324284 |