Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025570.2 | NP_079846.1 | 2 | 186 | ||
GenBank mRNA | AB049645.1 | - | 2 | 159 | ||
AK004053.1 | - | 2 | 171 | |||
AK011341.1 | - | 2 | 187 | |||
AK015199.1 | - | 170 | ||||
AK168665.1 | - | 2 | 160 | |||
BC016591.1 | - | 2 | 149 | 3158162 | ||
BC039985.1 | - | 2 | 156 | 1244957 |