Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025575.3 | NP_079851.2 | 4 | 939 | ||
GenBank mRNA | AK003071.1 | - | 4 | 932 | ||
AK005249.1 | - | 4 | 842 | |||
AK005510.1 | - | 4 | 876 | |||
AK007882.1 | - | 4 | 938 | |||
AK009684.1 | - | 4 | 903 | |||
AK011753.1 | - | 4 | 935 | |||
AK013220.1 | - | 4 | 912 | |||
AK017524.1 | - | 3 | 771 | |||
AK032098.1 | - | 3 | 774 | |||
AK075770.1 | - | 4 | 904 | |||
AK148305.1 | - | 4 | 877 | |||
AK210170.1 | - | 1 | 293 | |||
AK216375.1 | - | 1 | 297 | |||
AK219257.1 | - | 1 | 297 | |||
BC053943.1 | - | 3 | 773 | |||
BC061084.1 | - | 3 | 813 | 6596164 |