Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025576.2 | NP_079852.1 | 3 | 596 | ||
GenBank mRNA | AK003210.1 | - | 3 | 300 | ||
AK012674.1 | - | 3 | 586 | |||
AK014691.1 | - | 3 | 590 | |||
AK151972.1 | - | 3 | 597 | |||
AK168416.1 | - | 462 | ||||
BC026750.1 | - | 3 | 575 | 5135841 |