Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025576.2 | NP_079852.1 | 4 | 757 | ||
GenBank mRNA | AK003210.1 | - | 4 | 461 | ||
AK012674.1 | - | 4 | 747 | |||
AK014691.1 | - | 4 | 751 | |||
AK151972.1 | - | 4 | 758 | |||
AK168416.1 | - | 623 | ||||
BC026750.1 | - | 4 | 736 | 5135841 |