Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810428I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025577.2 | NP_079853.1 | 5 | 499 | ||
GenBank mRNA | AK010635.1 | - | 5 | 496 | ||
AK013181.1 | - | 5 | 500 | |||
AK160511.1 | - | 5 | 443 | |||
BC028495.1 | - | 5 | 505 | 3370477 |