Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025578.4 | NP_079854.2 | 4 | 1397 | ||
GenBank mRNA | AK013196.1 | - | 4 | 1355 | ||
AK131114.1 | - | 4 | 1326 | |||
BC022953.1 | - | 4 | 1384 | 5126320 |