Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172500.1 | - | 4, 5 | 516 | ||
BC019940.1 | - | 4, 5 | 464 | 3709649 | ||
DQ926863.1 | - | 4, 5 | 491 | |||
DQ926864.1 | - | 4, 5 | 555 |