Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025588.2 | NP_079864.1 | 3 | 376 | ||
GenBank mRNA | AK016532.1 | - | 4 | 503 | ||
AK032766.1 | - | 3 | 398 | |||
AK042744.1 | - | 3 | 423 | |||
AK050068.1 | - | 3 | 384 | |||
AK180355.1 | - | 2 | 269 | |||
BC048154.1 | - | 3 | 376 | 6400932 | ||
BC049102.1 | - | 3 | 376 | 5702581 |