Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025607.3 | NP_079883.3 | 2 | 180 | ||
GenBank mRNA | AK010854.1 | - | 2 | 193 | ||
AK018088.1 | - | 1 | 625 | |||
AK018849.1 | - | 3 | 301 | |||
AK159788.1 | - | 2 | 193 | |||
AK166904.1 | - | 2 | 192 | |||
AK166995.1 | - | 2 | 193 | |||
AK167396.1 | - | 2 | 193 | |||
AK167633.1 | - | 2 | 193 | |||
AK167935.1 | - | 2 | 193 | |||
AK168670.1 | - | 2 | 194 | |||
BC005559.1 | - | 2 | 204 | |||
BC083119.1 | - | 2 | 181 | 5374213 |