Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025613.3 | NP_079889.2 | 1 | 490 | ||
GenBank mRNA | AF509968.1 | - | 1 | 428 | ||
AK002559.1 | - | 1 | 496 | |||
AK005258.1 | - | 1 | 460 | |||
AK011293.1 | - | 1 | 482 | |||
AK012453.1 | - | 1 | 482 | |||
AK014440.1 | - | 1 | 436 | |||
AK017543.1 | - | 1 | 466 | |||
AK077783.1 | - | 1 | 470 | |||
AK078402.1 | - | 1 | 436 | |||
AK078409.1 | - | 1 | 436 | |||
AK078826.1 | - | 1 | 414 | |||
AK148555.1 | - | 1 | 431 | |||
AK169604.1 | - | 1 | 464 | |||
AK172612.1 | - | 1 | 271 | |||
AL357453.1 | - | 1 | 456 | |||
BC010712.1 | - | 1 | 475 | |||
BC069897.1 | - | 1 | 413 | |||
BY703074.1 | - | 1 | 488 |