Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025617.2 | NP_079893.1 | 5 | 622 | ||
GenBank mRNA | AK008715.1 | - | 5 | 577 | ||
AK011991.1 | - | 5 | 563 | |||
BC028321.1 | - | 5 | 560 | 3992258 |