Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntpcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025636.5 | NP_079912.1 | 4 | 530 | ||
GenBank mRNA | AK006433.1 | - | 4 | 438 | ||
AK010229.1 | - | 4 | 505 | |||
AK010823.1 | - | 4 | 531 | |||
AK019321.1 | - | 4 | 510 | |||
AK019421.1 | - | 4 | 501 | |||
BC025168.1 | - | 4 | 438 | 4223622 |