Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025638.2 | NP_079914.1 | 9 | 955 | ||
GenBank mRNA | AK011487.1 | - | 9 | 878 | ||
AK013864.1 | - | 9 | 881 | |||
AK016023.1 | - | 9 | 881 | |||
AK167812.1 | - | 9 | 938 | |||
BC016541.1 | - | 9 | 878 | 3602449 |