Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025638.2 | NP_079914.1 | 10 | 2147 | ||
GenBank mRNA | AK039183.1 | - | 1 | 1129 | ||
AK167812.1 | - | 10 | 2126 | |||
AK202723.1 | - | 1 | 72 | |||
BC016541.1 | - | 10 | 2072 | 3602449 | ||
CA451251.1 | - | 1 | 452 |